Merck
  • Urine glyceraldehyde excretion is elevated in the renal Fanconi syndrome.
Urine glyceraldehyde excretion is elevated in the renal Fanconi syndrome.
Kidney international (1989-01-01)
A J Jonas, S N Lin, S B Conley, J A Schneider, J C Williams, R C Caprioli
ABSTRACT
We analyzed urinary constituents using GC/MS in 16 children with the renal Fanconi syndrome and 13 normal individuals. Urine glyceraldehyde levels were strikingly elevated in the renal Fanconi syndrome group (mean 5.1 +/- 4.8 mg/mg creatinine) compared to levels in the normal group (mean 0.04 +/- 0.04 mg/mg creatinine, P less than 0.001). Urine lactate levels were also elevated in the renal Fanconi syndrome group (mean 2.3 +/- 2.6 mg/mg creatinine) compared to normals (mean 0.01 +/- 0.01 mg/mg creatinine, P less than 0.003). Only small elevations of glyceraldehyde and lactate were found in urine from children with other renal disorders. Serum levels of glyceraldehyde and lactate were no greater in individuals with the Fanconi syndrome than in the normals. The fractional reabsorption of both glyceraldehyde and lactate was virtually complete in the normals, but was markedly impaired in the Fanconi syndrome patients where, in some cases, glyceraldehyde excretion greatly exceeded the excretion of creatinine. We conclude that marked glyceraldehyde excretion is a previously unrecognized feature of the renal Fanconi syndrome which may result from disordered proximal tubular glycolytic metabolism. Further studies will be required to determine the role of glyceraldehyde loss in the pathogenesis of this generalized disturbance of proximal tubular function.
Sigma-Aldrich
L-(+)-Lactic acid, 80%
Sigma-Aldrich
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%
Sigma-Aldrich
L-(+)-Lactic acid, Vetec, reagent grade, 86%
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O