Merck
  • Appearance of early and late components of Epstein-Barr virus-associated membrane antigen in Daudi cells superinfected with P3HR-1 virus.
Appearance of early and late components of Epstein-Barr virus-associated membrane antigen in Daudi cells superinfected with P3HR-1 virus.
The Journal of general virology (1978-01-01)
T Sairenji, Y Hinuma, T Sekizawa, M Yoshida
PMID202665
ABSTRACT
This study investigated the synthesis of membrane antigen (MA) as well as virus capsid antigen (VCA) and early antigen (EA) in Daudi cells which had been superinfected with the P3HR-1 strain of Epstein-Barr virus (EBV) and then treated with trypsin to remove initially absorbed MA-positive material from the cell surface. Synthesis of MA, VCA and EA was completely inhibited by puromycin. A marked reduction in the frequency of MA positive cells was observed in superinfected cells cultured in the presence of either cytosine arabinoside (Ara-C) or phosphonoacetate (PA); however, a small fraction of MA synthesis occurred, suggesting an inhibitor insensitive component in MA, A differential absorption of EBV antibody-positive human serum with the Ara-C treated or untreated infected cells detected two antigenically different components in MA: early (Ara-C insensitive) and late (Ara-C sensitive) MA.
Sigma-Aldrich
Phosphonoacetic acid, 98%
Pricing and availability is not currently available.