Merck
  • Inflammatory multiple-sclerosis plaques generate characteristic metabolic profiles in cerebrospinal fluid.
Inflammatory multiple-sclerosis plaques generate characteristic metabolic profiles in cerebrospinal fluid.
PloS one (2007-07-06)
Norbert W Lutz, Angèle Viola, Irina Malikova, Sylviane Confort-Gouny, Bertrand Audoin, Jean-Philippe Ranjeva, Jean Pelletier, Patrick J Cozzone
ABSTRACT
Multiple sclerosis (MS), an inflammatory disease of the central nervous system, manifests itself in numerous forms and stages. A number of brain metabolic alterations have been reported for MS patients vs. control subjects. However, metabolite profiles of cerebrospinal fluid (CSF) are not consistent among the published MS studies, most probably due to variations in the patient cohorts studied. We undertook the first investigation of highly homogeneous MS patient cohorts to determine characteristic effects of inflammatory MS plaques on the CSF metabolome, including only patients with clinically isolated syndrome (CIS) with or without inflammatory brain plaques, and controls. CSF obtained by lumbar puncture was analyzed by proton magnetic resonance spectroscopy. 27 metabolites were quantified. Differences between groups of control subjects (n = 10), CIS patients with (n = 21) and without (n = 12) inflammatory plaques were evaluated by univariate statistics and principal component analysis (PCA). Seven metabolites showed statistically significant inter-group differences (p<0.05). Interestingly, a significant increase in beta-hydroxyisobutyrate (BHIB) was detected in CIS with vs. without active plaques, but not when comparing either CIS group with control subjects. Moreover, a significant correlation was found, for the first time, between CSF lactate concentration and the number of inflammatory MS brain plaques. In contrast, fructose concentrations were equally enhanced in CIS with or without active plaques. PCA based on all 27 metabolites yielded group-specific clusters. CSF metabolic profiles suggest a close link between MS plaque activity in CIS patients on the one hand and organic-acid metabolism on the other. Our detection of increased BHIB levels points to a hitherto unsuspected role for this compound in MS with active plaques, and serves as a basis for further investigation. The metabolic effects described in our study are crucial elements in the explanation of biochemical mechanisms involved in specific MS manifestations.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Coenzyme Q1, ≥95%
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol