Merck
  • Serum hyaluronan concentration as a marker of angiopathy in patients with diabetes mellitus.
Serum hyaluronan concentration as a marker of angiopathy in patients with diabetes mellitus.
Endocrine journal (2006-09-13)
Shinichiro Mine, Yosuke Okada, Chie Kawahara, Takahiro Tabata, Yoshiya Tanaka
ABSTRACT
Accumulation of hyaluronan (HA) around smooth muscle cells in lesions of atherosclerosis in diabetic patients suggests that this protein plays an important role in diabetic angiopathy. The aim of this study was to determine the correlation between serum HA concentrations and diabetic angiopathy. Diabetic patients treated with or without an oral hypoglycemic agent and/or insulin for at least 1 year were recruited (n = 95). We also included 20 non-diabetic control subjects. We measured serum levels of HA, body mass index (BMI), fasting plasma glucose (FPG), HbA1c, total cholesterol, triglyceride, glycated albumin (GA), high sensitivity CRP (hs-CRP), monocyte chemoattractant protein (MCP)-1 and evaluated diabetes mellitus history, drug use and presence of related complications. Serum HA levels were significantly (P<0.05) higher in diabetic patients (83.6 +/- 5.6 ng/ml, mean +/- SEM) than in normal subjects (41.7 +/- 12 ng/ml). In diabetic patients, serum HA concentration significantly correlated with FPG, HbA1c, GA, triglyceride and also significantly correlated with BMI, hs-CRP and MCP-1 and tended to be higher in diabetic patients with complications than in those without such complications. Our data suggest that serum HA level correlates with poor blood glucose control and diabetic angiopathy and that it could be used as a marker of diabetic angiopathy.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Supelco
1-Butanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%