Merck
  • Effect of oral creatine supplementation on urinary methylamine, formaldehyde, and formate.
Effect of oral creatine supplementation on urinary methylamine, formaldehyde, and formate.
Medicine and science in sports and exercise (2005-11-02)
Jacques R Poortmans, Alain Kumps, Pierre Duez, Aline Fofonka, Alain Carpentier, Marc Francaux
ABSTRACT
It has been claimed that oral creatine supplementation might have potential cytotoxic effects on healthy consumers by increasing the production of methylamine and formaldehyde. Despite this allegation, there has been no scientific evidence obtained in humans to sustain or disprove such a detrimental effect of this widely used ergogenic substance. Twenty young healthy men ingested 21 g of creatine monohydrate daily for 14 consecutive days. Venous blood samples and 24-h urine were collected before and after the 14th day of supplementation. Creatine and creatinine were analyzed in plasma and urine, and methylamine, formaldehyde, and formate were determined in 24-h urine samples. Oral creatine supplementation increased plasma creatine content 7.2-fold (P < 0.001) and urine output 141-fold (P < 0.001) with no effect on creatinine levels. Twenty-four-hour urine excretion of methylamine and formaldehyde increased, respectively, 9.2-fold (P = 0.001) and 4.5-fold (P = 0.002) after creatine feeding, with no increase in urinary albumin output (9.78 +/- 1.93 mg x 24 h(-1) before, 6.97 +/- 1.15 mg x 24 h(-1) creatine feeding). This investigation shows that short-term, high-dose oral creatine supplementation enhances the excretion of potential cytotoxic compounds, but does not have any detrimental effects on kidney permeability. This provides indirect evidence of the absence of microangiopathy in renal glomeruli.
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Creatine, anhydrous