Merck
  • Homocysteine and methylmalonic acid concentrations in cerebrospinal fluid: relation with age and Alzheimer's disease.
Homocysteine and methylmalonic acid concentrations in cerebrospinal fluid: relation with age and Alzheimer's disease.
Journal of neurology, neurosurgery, and psychiatry (2005-10-18)
J-M Serot, F Barbé, E Arning, T Bottiglieri, P Franck, P Montagne, J-P Nicolas
ABSTRACT
Homocysteine may be involved in the pathogenesis of late onset Alzheimer's disease. It is implicated in the metabolism of several important pathways in the brain. Methylmalonic acid (MMA) is related to the metabolism of branched chained amino acids and fatty acids. To compare cerebrospinal fluid (CSF) total homocysteine and MMA in elderly subjects, patients with Alzheimer's disease, and younger healthy controls. CSF samples were obtained from 33 patients under 20 years of age; 28 patients aged 21 to 60 years; 22 normal elderly subjects aged over 60; and 38 Alzheimer patients aged over 60. CSF total homocysteine increased with age (mean (SD): 57 (35) nmol/l in the youngest group v 123 (89) nmol/l in the elderly group (p<0.001)). There was no difference between the elderly group and Alzheimer patients (115 (62) nmol/l). CSF MMA did not differ in the elderly group and the Alzheimer group (38 (13) v 35 (14) ng/ml). In the youngest group, it was significantly higher (60 (15) ng/ml). CSF total homocysteine is not increased in Alzheimer's disease compared with age matched controls. CSF total homocysteine was correlated with age. The decrease in CSF MMA levels with age eliminates a lack of vitamin B-12 at neuronal level.
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%