Merck
Molybdenum-containing hydroxylases.
Archives of biochemistry and biophysics (2004-12-08)
Russ Hille
ABSTRACT
Unlike monooxygenases, molybdenum-containing hydroxylases catalyze the hydroxylation of carbon centers using oxygen derived ultimately from water, rather than O(2), as the source of the oxygen atom incorporated into the product, and do not require an external source of reducing equivalents. The mechanism by which this interesting chemistry takes place has been the subject of investigation for some time, and in the last several years the chemical course of the reaction has become increasingly well understood. The present minireview summarizes recent mechanistic and structure/function studies of members of this large and growing family of enzymes.
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis