Merck
  • Levels of 7-oxocholesterol in cerebrospinal fluid are more than one thousand times lower than reported in multiple sclerosis.
Levels of 7-oxocholesterol in cerebrospinal fluid are more than one thousand times lower than reported in multiple sclerosis.
Journal of lipid research (2004-12-04)
Valerio Leoni, Dieter Lütjohann, Thomas Masterman
ABSTRACT
In a recent publication [Diestel, A., O. Aktas, D. Hackel, I. Hake, S. Meier, C. S. Raine, R. Nitsch, F. Zipp, and O. Ullrich. 2003. Activation of microglial poly (ADP-ribose)-polymerase-1 by cholesterol breakdown products during neuroinflammation: a link between demyelination and neuronal damage. J. Exp. Med. 198: 1729-1740], extremely high levels of 7-oxocholesterol were reported in cerebrospinal fluid (CSF) of 11 patients with multiple sclerosis (MS) [7.4 +/- 0.3 mg/l (mean +/- SEM)]. The corresponding level of 12 subjects with other kinds of neurological diseases was reported to be 0.5 +/- 0.1 mg/l. Such high levels of 7-oxocholesterol were found to cause neuronal damage of living brain tissues. Using a highly accurate method for an assay of 7-oxocholesterol based on isotope dilution-mass spectrometry and anaerobic conditions during workup, we found that the level of 7-oxocholesterol in CSF from 29 Swedish patients with MS was only 1.2 microg/l (median, ranging from 0.4 to 4.6 microg/l), less than 1/1,000 th of the previously reported level. The level of 7-oxocholesterol in CSF from 24 Swedish control patients was 0.9 microg/l (0.3-2.3 microg/l), slightly but significantly lower than the CSF level in MS patients (P=0.002). In vitro-induced lipid peroxidation of the endogenous cholesterol in CSF increased the level of 7-oxygenated cholesterol metabolites, particularly 7-oxocholesterol, up to approximately 0.3 mg/l. These results are discussed in relation to the fact that 7-oxygenated steroids are easily artificially formed by autoxidation of cholesterol during workup procedures and analysis of sterols and oxysterols from biological samples.
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Coenzyme Q1, ≥95%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis