Merck
Semicarbazide-sensitive amine oxidases: enzymes with quite a lot to do.
Neurotoxicology (2003-12-31)
Jeff O'Sullivan, Mercedes Unzeta, Joe Healy, Michael I O'Sullivan, Gavin Davey, Keith F Tipton
ABSTRACT
The semicarbazide-sensitive amine oxidases (SSAO) (EC 1.4.3.6) were believed to be detoxifying enzymes, primarily involved in the oxidative deamination of endogenous amines, such as methylamine and aminoacetone, together with some xenobiotic amines. However, it appears that the reaction products may have important signalling functions in the regulation of cell development and glucose homeostasis. Furthermore, enzyme, from some sources, behaves as a cellular adhesion protein under inflammatory and it may also be involved in lipid transport. This review considers what is known about the activities and potential functions of this hardworking protein.
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol