Merck
  • [Isopropanol and acetone level in serum after preoperative surface disinfection with antiseptics containing isopropanol].
[Isopropanol and acetone level in serum after preoperative surface disinfection with antiseptics containing isopropanol].
Blutalkohol (1992-09-01)
S Wittmann, T Gilg, H G Dietz, R Grantzow, O Peschel, L von Meyer
ABSTRACT
After preoperative skin disinfection in pediatric surgery, serum levels of isopropanol up to 12.2 mg/l (MW 5.0 mg/l +/- 3.37, n = 26) were found. They result from a rapid and prolonged but uncharacteristic percutaneous resorption of the isopropanol-containing disinfectant. In about 50% of the cases, serum levels of acetone showed an increase up to 82 mg/l already before skin disinfection, presumably caused by preoperative starvation. After skin disinfection, raised acetone levels were found in 19 of 26 cases. As increased isopropanol and acetone levels are discussed as alcoholism markers, a falsification of congener analysis after skin disinfection, e.g. in cases of adult victims of accidents, has to be taken into consideration. Endogenous serum levels of methanol (0.87 mg/l +/- 0.49), ethanol (0.32 mg/l +/- 0.09), acetaldehyde (0.31 mg/l +/- 0.10) and others remained unaffected. Some uncharacteristic elevations of propanol-1 levels are caused by contaminated rubber caps.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Acetone, analytical standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Isopropanol, 70% in H2O
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)