Merck
  • Pentosidine and N(epsilon)-(carboxymethyl)-lysine in Alzheimer's disease and vascular dementia.
Pentosidine and N(epsilon)-(carboxymethyl)-lysine in Alzheimer's disease and vascular dementia.
Neurobiology of aging (2002-12-25)
K J Bär, S Franke, B Wenda, S Müller, R Kientsch-Engel, G Stein, H Sauer
ABSTRACT
Increasing evidence suggests an interaction of oxidative stress and the formation of advanced glycation end products (AGE) in the onset and progression of Alzheimer's disease. We studied levels of pentosidine and N(epsilon)-(carboxymethyl)-lysine (CML) in serum and cerebrospinal fluid (CSF) of 15 patients with probable Alzheimer's disease (AD), 20 patients with vascular dementia (VD), and 31 control subjects (14 matched for age, and 17 younger patients). AGE protein concentrations in CSF did not differ within controls when divided into two subgroups by age. We found significantly elevated levels of CML in CSF of AD patients and of pentosidine in CSF of patients suffering from vascular dementia when compared to controls. The concentrations of pentosidine and CML in serum apparently did not relate directly to CSF values, suggesting influence of extra-cerebral factors in serum samples. It is concluded that AGE proteins are differentially affected in these types of dementia, depending on the specific neuropathology. Furthermore, measurements of AGE products in vivo should rely on CSF rather than blood samples.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol