Merck
  • Variation in polyol levels in cerebrospinal fluid and serum in diabetic patients.
Variation in polyol levels in cerebrospinal fluid and serum in diabetic patients.
Diabetologia (1975-12-01)
C Servo, E Pitkänen
ABSTRACT
Cerebrospinal fluid (CSF) or CSF and plasma levels of sorbitol, 1,5-anhydroglucitol and myoinositol of diabetic and non-diabetic patients with normal kidney function and of diabetic and non-diabetic patients with impaired kidney function were measured by gas-liquid chromatography. The CSF sorbitol level correlated with the plasma glucose level (p less than or equal to 0.05) in diabetic patients with normal kidney function, having received insulin for less than 12 months. The correlation between CSF sorbitol and plasma glucose levels in patients not dependent on insulin was not significant. Sorbitol was not detected in the plasma. The highest sorbitol levels in CSF were seen in insulin-dependent diabetic patients with impaired kidney function. No rise was seen in non-diabetic uremia. 1,5-anhydroglucitol, normally present in plasma, was absent from CSF and plasma in diabetic patients receiving insulin. In non-diabetic uremic patients, 1,5-anhydroglucitol levels in CSF and plasma were lower than in healthy subjects, but htere was no correlation with plasma glucose levels. The myoinositol level was higher in CSF than in the plasma of both non-diabetic and diabetic patients with normal kidney function. Both plasma and CSF levels were significantly (p less than 0.001) elevated in diabetic as well as in non-diabetic uremic patients, the plasma myoinositol increasing relatively more than the CSF levels. The elevation of plasma myoinositol correlated with the elevation of plasma creatinine and thus also with the impairment of kidney function. Plasma and CSF myoinositol levels were not influenced by the plasma glucose level.
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Sigma-Aldrich
myo-Inositol, Vetec, reagent grade, 99%
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology