Merck
  • Increased levels of the pro-inflammatory prostaglandin PGE2 in CSF from ALS patients.
Increased levels of the pro-inflammatory prostaglandin PGE2 in CSF from ALS patients.
Neurology (2002-04-24)
G Almer, P Teismann, Z Stevic, J Halaschek-Wiener, L Deecke, V Kostic, S Przedborski
ABSTRACT
Levels of the potent pro-inflammatory prostaglandin E(2) (PGE(2)) are elevated in postmortem spinal cords from patients with ALS, and inhibition of a key PGE(2)-synthesizing enzyme, cylcooxygenase-2, is neuroprotective in an in vitro model of ALS. The authors report that 82% of the patients with ALS studied had 2 to 10 times higher PGE(2) levels in CSF compared with normal control subjects. That affected areas of the CNS are inflamed in ALS supports this. CSF PGE(2) measurement may be useful in monitoring treatment for ALS.
Product Description
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture