Merck
  • High-performance liquid chromatographic determination of ursodeoxycholic acid after solid phase extraction of blood serum and detection-oriented derivatization.
High-performance liquid chromatographic determination of ursodeoxycholic acid after solid phase extraction of blood serum and detection-oriented derivatization.
Journal of pharmaceutical and biomedical analysis (2001-03-15)
M Nobilis, M Pour, J Kunes, J Kopecký, J Kvĕtina, Z Svoboda, K Sládková, J Vortel
ABSTRACT
Ursodeoxycholic acid (3 alpha,7 beta-dihydroxy-5 beta-cholanoic acid, UDCA) is a therapeutically applicable bile acid widely used for the dissolution of cholesterol-rich gallstones and in the treatment of chronic liver diseases associated with cholestasis. UDCA is more hydrophilic and less toxic than another therapeutically valuable bile acid, chenodeoxycholic acid (CDCA), the 7 alpha-epimer of UDCA. Procedures for sample preparation and HPLC determination of UDCA in blood serum were developed and validated. A higher homologue of UDCA containing an additional methylene group in the side chain was synthetized and used as an internal standard (IS). Serum samples with IS were diluted with a buffer (pH=7). The bile acids and IS were captured using solid phase extraction (C18 cartridges). The carboxylic group of the analytes was derivatized using 2-bromo-2'-acetonaphthone (a detection-oriented derivatization), and reaction mixtures were analyzed (HPLC with UV 245 nm detection; a 125--4 mm column containing Lichrospher 100 C18, 5 microm; mobile phase: acetonitrile--water, 6:4 (v/v)). Following validation, this method was used for pharmacokinetic studies of UDCA in humans.
Product Description
Supelco
Supelclean LC-18 SPE Tube, bed wt. 100 mg, volume 1 mL, pk of 108
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard