Merck
Chemical composition of some wild peanut species (Arachis L.) seeds.
Journal of agricultural and food chemistry (2000-03-22)
N R Grosso, V Nepote, C A Guzmán
ABSTRACT
Oil, protein, ash, and carbohydrate contents, iodine value, and fatty acid and sterol compositions were studied in seeds of Arachis trinitensis, A. chiquitana, A. kempff-mercadoi, A. diogoi, A. benensis, A. appressipila, A. valida, A. kretschmeri, A. helodes, A. kuhlmannii, A. williamsii, A. sylvestris, A. matiensis, A. pintoi, A. hoehnei, A. villosa, and A. stenosperma. Oil content was greatest in A.stenosperma (mean value = 51.8%). The protein level was higher in A. sylvestris (30.1%) and A. villosa (29.5%). Mean value of oleic acid varied between 30.6% (A. matiensis) and 46.8% (Arachis villosa), and linoleic acid oscillated between 34.1% (A. villosa) and 47.4% (A. appressipila). The better oleic-to-linoleic (O/L) ratio was exhibited by A. villosa (1.38). Some species showed higher concentration of behenic acid. The greatest level of this fatty acid was found in A. matiensis (6.2%). Iodine value was lower in A. valida (99.2). The sterol composition in the different peanut species showed higher concentration of beta-sitosterol (mean values oscillated between 55.7 and 60.2%) followed by campesterol (12.4-16. 5%), stigmasterol (9.7-13.3%), and Delta(5)-avenasterol (9.7-13.4%). The chemical quality and stability of oils (iodine value and O/L ratio) from wild peanut studied in this work are not better than those of cultivated peanut.
Sigma-Aldrich
Behenic acid, 99%
Supelco
Behenic acid, analytical standard