Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Alpha-cyano-4-hydroxycinnamic acid affinity sample preparation. A protocol for MALDI-MS peptide analysis in proteomics.
Alpha-cyano-4-hydroxycinnamic acid affinity sample preparation. A protocol for MALDI-MS peptide analysis in proteomics.
Analytical chemistry (2001-02-24)
J Gobom, M Schuerenberg, M Mueller, D Theiss, H Lehrach, E Nordhoff
ABSTRACT
We present a new MALD1 sample preparation technique for peptide analysis using the matrix alpha-cyano-4-hydroxy-cinnamic acid (CHCA) and prestructured sample supports. The preparation integrates sample purification, based on the affinity of microcrystalline CHCA for peptides, thereby simplifying the analysis of crude peptide mixtures. Enzymatic digests can thus be prepared directly, without preceding purification. Prepared samples are homogeneous, facilitating automatic spectra acquisition. This method allows preparation of large numbers of samples with little effort and without the need for automation. These features make the described preparation suitable for cost-efficient high-throughput protein identification. Performance of the sample preparation is demonstrated with in situ proteolytic digests of human brain proteins separated by two-dimensional gel electrophoresis.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)