Merck
  • Simultaneous multi-lifetime multi-color STED imaging for colocalization analyses.
Simultaneous multi-lifetime multi-color STED imaging for colocalization analyses.
Optics express (2011-03-04)
Johanna Bückers, Dominik Wildanger, Giuseppe Vicidomini, Lars Kastrup, Stefan W Hell
ABSTRACT
We describe a STED microscope optimized for colocalization experiments with up to three colors. Two fluorescence labels are separated by their fluorescence lifetime whereas a third channel is discriminated by the wavelength of fluorescence emission. Since it does not require a second STED beam, separating by lifetime is insensitive to drift and thus optimally suited for colocalization analyses. Furthermore, we propose a setup having a second STED beam for long duration multicolor recording.
Sigma-Aldrich
Phalloidin–Atto 532, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 647N amine, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N azide, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
Atto 425-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N alkyne, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 390 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N DOPE, suitable for fluorescence
Sigma-Aldrich
Atto 647N-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 647N-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 425, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N Protein Labeling Kit, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 590-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 590, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 590 NHS ester, BioReagent, suitable for fluorescence, ≥90.0% (degree of coupling)
Supelco
Atto 647N, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Supelco
Atto 390 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Supelco
NTA - Atto 647 N, BioReagent, suitable for fluorescence, ≥80% (HPCE)
Sigma-Aldrich
Atto 590-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Abberior® STAR RED NHS, ≥80.0% (degree of coupling)
SAFC
Atto 465 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)