Sigma-Aldrich
MAPTrix™ Adhesive, Low Molecular Weight, Tyrosinase-Pretreated, lyophilized powder
Sigma-Aldrich
MAPTRIX-E-LRAHAVDING, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTrix™ Adhesive, High Molecular Weight, Tyrosinase-Pretreated, lyophilized powder
Sigma-Aldrich
MAPTRIX-E-LRAHAVDING, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-V-KKQRFRHRNRKGY, Vitronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-GLPGER, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-M-VAEIDGIEL, Tenascin-C Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F- PHSRN, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-V-FRHRNRKGY, Vitronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-NRWHSIYITRFG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-YIGSR, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTrix™ Reagent, Low Molecular Weight, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RQVFQVAYIIIKA, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-GLPGER, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-KNSFMALYLSKG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTrix™ Reagent, High Molecular Weight, aqueous solution
Sigma-Aldrich
MAPTrix™ Reagent, High Molecular Weight, aqueous solution
Sigma-Aldrich
MAPTRIX-F-RGD, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C- GFPGER, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RYVVLPR, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-V-FRHRNRKGY, Vitronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-IKVAV, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTrix™ Reagent, Low Molecular Weight, lyophilized powder
Sigma-Aldrich
MAPTRIX-L-IKVAV, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-E-ADTPPV, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-TWYKIAFQRNRK, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RNIAEIIKDI, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F-REDV, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-TWYKIAFQRNRK, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F-PHSRN-RGDSP, Fibronectin Mimetic, aqueous solution