Merck
  • Review of recent placebo-controlled trials utilizing phytotherapeutic agents for treatment of BPH.
Review of recent placebo-controlled trials utilizing phytotherapeutic agents for treatment of BPH.
The Prostate (1998-10-29)
F C Lowe, K Dreikorn, A Borkowski, J Braeckman, L Denis, P Ferrari, G Gerber, R Levin, P Perrin, T Senge
ABSTRACT
In order to assess the efficacy of phytotherapeutic agents for the treatment of benign prostatic hyperplasia (BPH), a review of recently published double-blind placebo-controlled trials was undertaken. Only those studies reviewed by the Other Medical Therapies Committee of the Fourth International Consultation on BPH were included. These studies suggest a possible benefit for the use of phytotherapeutic preparations in the treatment of BPH. These studies need to be confirmed in larger long-term placebo-controlled studies in order to ascertain the true efficacy of these agents.