Merck
  • Vitamins and monothiols efficacy in the restoration of adenosine nucleotide degradation enzymes altered during methylmercury intoxication.
Vitamins and monothiols efficacy in the restoration of adenosine nucleotide degradation enzymes altered during methylmercury intoxication.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (1995-01-01)
P P Sood, C Bapu, K Vijayalakshmi
ABSTRACT
Male albino mice were intoxicated with a daily dose of 1 mg/kg of methylmercury chloride (MMC) for 7 days, and were treated thereafter with glutathione, N-acetyl-DL-homocysteine thiolactone, vitamin B complex, and vitamin E, either alone or in combinations, for the next 7 days. The animals were sacrificed on the eighth day, with the exception of one group that was kept without toxic exposure for an additional 7 days and sacrificed on the fifteenth day. Brain, spinal cord, kidney, and liver of the animals were examined for changes in adenosine deaminase and 5' nucleotidase. We found a severe inhibition of these enzymes during MMC intoxication and significant recovery during monothiols and vitamins administration, indicating the effectiveness of these agents in methylmercury detoxication.
Sigma-Aldrich
DL-N-Acetylhomocysteine thiolactone, 98%