Merck
  • Determination of the pesticides diflubenzuron and clofentezine in plums, strawberries and blackcurrant-based fruit drinks by high performance liquid chromatographic/atmospheric pressure chemical ionization-mass spectrometry.
Determination of the pesticides diflubenzuron and clofentezine in plums, strawberries and blackcurrant-based fruit drinks by high performance liquid chromatographic/atmospheric pressure chemical ionization-mass spectrometry.
Rapid communications in mass spectrometry : RCM (1995-01-01)
K A Barnes, R J Fussell, J R Startin, S A Thorpe, S L Reynolds
ABSTRACT
A high performance liquid chromatographic/atmospheric pressure chemical ionization-mass spectrometric (HPLC/APCI-MS) method has been developed for the determination of the pesticides diflubenzuron (1-(4-chlorophenyl)-3-(2,6-difluorobenzoyl)urea) and clofentezine (3,6-bis(2-chlorophenyl)-1,2,4,5-tetrazine) in plums, strawberries and blackcurrant-based fruit drinks. Samples were homogenized with acetone, extracted into dichloromethane + cyclohexane and cleaned-up by high performance gel permeation chromatography. HPLC was performed on an ODS column with methanol + water at 1 mL/min. Detection was by negative-ion selected-ion monitoring APCI-MS. Comparison of response with solvent and matrix-matched standards showed some enhancement of response for the latter, and these standards were consequently used for quantification. The calibration was linear over the range 0.05-0.50 ng/microL in all three matrices. The mean overall recovery of diflubenzuron and clofentezine from spiked extracts (0.086 mg/kg) in all three matrices was 76% and 70% respectively with relative standard deviations of 15% and 12% respectively (n = 12). The limit of detection was both compound and commodity dependent and ranged from 0.01-0.05 ng/microL, equivalent to 0.003-0.014 mg/kg in the crop.
Product Description
Supelco
Clofentezine, PESTANAL®, analytical standard