- The conformation of the monomethyl ethers of methyl beta-lactoside in D2O and Me2SO-d6 solutions.
The conformation of the monomethyl ethers of methyl beta-lactoside in D2O and Me2SO-d6 solutions.
Carbohydrate research (1993-10-04)
P Fernández, J Jiménez-Barbero
PMID8252531
ABSTRACT
The solution conformations of all the possible monomethyl ethers of methyl beta-lactoside have been analysed using molecular mechanics and dynamics calculations and nuclear magnetic resonance data (variable temperature and NOE experiments). The overall shape of all the compounds studied is fairly similar and may be described by conformers included in a low-energy region with phi = -100 +/- 40 degrees and psi = -135 +/- 35 degrees, which is ca. 5% of the total potential energy surface for the glycosidic linkages of the disaccharides.