Merck
  • Application of polydimethylsiloxane rod extraction to the determination of sixteen halogenated flame retardants in water samples.
Application of polydimethylsiloxane rod extraction to the determination of sixteen halogenated flame retardants in water samples.
Analytica chimica acta (2013-03-19)
Carme Valls-Cantenys, Eugenia Villaverde-de-Sáa, Rosario Rodil, José Benito Quintana, Mònica Iglesias, Victòria Salvadó, Rafael Cela
ABSTRACT
An extraction and preconcentration procedure for the determination in water samples of several halogenated flame retardants (FRs), nine brominated diphenyls ethers (BDEs) and seven non-BDE FRs, was developed and validated. The optimised procedure is based on polydimethylsiloxane (PDMS) rods as sorptive extraction material, followed by liquid desorption and gas chromatography coupled to negative chemical ionisation-mass spectrometry (GC-NCI-MS) determination, rendering an efficient and inexpensive method. The final optimised protocol consists of overnight extraction of 100mL of sample solutions containing 40% MeOH and 4% NaCl, followed by a 15-min sonication-assisted desorption with 300 μL of ethyl acetate, solvent evaporation and GC-NCI-MS analysis. Under these conditions, extraction efficiencies in the 9 to 70% range were obtained, leading to enrichment factors between 108 and 840, detection limits in the range from 0.4 to 10 ng L(-1)and RSD values in the 2-23% range. After method validation, different real water samples, including river, ria, sea, landfill leachate, influent and effluent wastewater from an urban sewage treatment plant (STP) and effluent wastewater from a textile industry, were analysed. BDE-47, BDE-99, BDE-100 and BDE-197 were detected in wastewater and landfill leachate samples at concentration levels up to 2887 ng L(-1). Among the non-BDE FRs, bis (2-ethylhexyl)-3,4,5,6-tetrabromo-phthalate (DEHTBP) was detected in surface water samples (sea, river and ria) between 1.3 and 2.2 ng L(-1) and 1,2-bis(2,4,6-tribromophenoxy)ethane (BTBPE) in the landfill leachate (64 ng L(-1)).
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses