Merck
  • Effects of estradiol benzoate and trenbolone acetate, alone or in combination at dose levels present in Synovex Choice, on performance by feedlot heifers.
Effects of estradiol benzoate and trenbolone acetate, alone or in combination at dose levels present in Synovex Choice, on performance by feedlot heifers.
Journal of animal science (2013-01-12)
R M Cleale, D Amodie, D T Bechtol, J S Drouillard, J D Edmonds, M Edmonds, B D Hunsaker, L A Kraft, T E Lawrence, R D Rulli, A R Waite
ABSTRACT
Trials were conducted with beef heifers at 4 sites to evaluate feedlot performance and carcass characteristics in response to implants containing 14 mg estradiol benzoate and 100 mg trenbolone acetate (EB/TBA; Synovex Choice, Zoetis LLC, New York, NY), 14 mg estradiol benzoate (EB), 100 mg trenbolone acetate (TBA), or a sham-implanted control (SC). The study design at each site was a randomized complete block with 12 blocks and 4 treatments. Blocks of cattle at each site were harvested in commercial abattoirs when masked personnel estimated at least 60% of animals would yield carcasses with USDA quality grades of Choice or Prime. Data were pooled across sites for statistical analysis. Initial BW averaged 374 kg, and days on feed ranged from 98 to 126 d (mean 112 d). Heifers implanted with EB/TBA, EB, and TBA had greater ADG and G:F (P < 0.05) than SC; ADG and G:F were greater for EB/TBA than EB or TBA (P < 0.05). Heifers treated with TBA had greater G:F than EB (P < 0.05). Feed intake was not affected by treatments. Mean HCW and LM area for EB/TBA were greater than for other treatments (P < 0.05). Mean HCW for TBA was greater than SC (P < 0.05) but not different from EB. Mean LM area for EB and TBA were greater than SC (P < 0.05) but not different from each other. There were no treatment differences (P > 0.05) for KPH, 12th-rib fat thickness, or yield grade. Dressing percent was greater for EB/TBA than SC (P < 0.05) but not different from EB or TBA. Marbling score was decreased by EB/TBA (P < 0.05) compared with other treatments, but no other differences were noted. Despite the effect of EB/TBA on marbling scores, there were no significant (P > 0.05) treatment differences on proportions of carcasses with quality grades ≥ Choice vs. < Choice. With respect to ADG and G:F, implants containing EB, TBA, or EB/TBA produced improved responses over SC. Furthermore, EB/TBA induced greater ADG and G:F responses than EB and TBA. Results confirmed that EB and TBA have additive effects, as evidenced by the observation that calves implanted with EB/TBA had significantly greater ADG and G:F than heifers implanted with either EB or TBA alone or compared with SC heifers.
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
Supelco
Trenbolone, analytical standard, for drug analysis