Merck
[Improved methodology for quantitative determination of thiabendazole].
Kokuritsu Iyakuhin Shokuhin Eisei Kenkyujo hokoku = Bulletin of National Institute of Health Sciences (2012-12-19)
Noriko Furusho, Noriko Otsuki, Takashi Ohtsuki, Chiye Tatebe-Sasaki, Kyoko Sato, Hiroshi Akiyama, Yoko Kawamura
ABSTRACT
The method prescribed in the 8th edition of Japan's Specifications and Standards for Food Additives (JSSFA) for the quantitative analysis of thiabendazole was improved by eliminating the use of toxic reagents such as mercuric acetate and chromium trioxide. For exclusion of mercuric acetate, a nonaqueous titration was performed using four types of solvent systems, including acetic acid:acetic anhydride (1:5), acetic acid:acetic anhydride (3:7), acetic acid alone, and formic acid:acetic acid (1:10), that did not contain mercuric compounds. Because precipitates were formed in titrations using acetic acid alone and formic acid:acetic acid (1:10), we considered that it was difficult to determine the purity using these solvent systems. However, it was confirmed that the purity of thiabendazole dissolved in the two acetic acid:acetic anhydride solvent systems can be determined using either a visual indicator or potentiometry. Specifically, the purity of thiabendazole was determined to be 99.9% (relative standard deviation (RSD) = 0.07%) for acetic acid:acetic anhydride (1:5) and 99.7% (RSD = 0.13%) for acetic acid:acetic anhydride (3:7) With respect to chromium trioxide, it was determined that chromium trioxide can be excluded using acetic acid, which conforms to the JIS K8001 standard for nonaqueous titrations. Therefore, in this study, an improved method for the quantitative determination of thiabendazole was developed without the use of toxic reagents.
Product Description
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
Tiabendazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Mercury(II) acetate, ACS reagent, ≥98.0%
Sigma-Aldrich
Mercury(II) acetate, puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)