Merck
  • Evaluation of the effect of hydroxypropyl-β-cyclodextrin on topical administration of milk thistle extract.
Evaluation of the effect of hydroxypropyl-β-cyclodextrin on topical administration of milk thistle extract.
Carbohydrate polymers (2012-12-12)
Gianpiera Spada, Elisabetta Gavini, Massimo Cossu, Giovanna Rassu, Antonio Carta, Paolo Giunchedi
ABSTRACT
Two water in oil emulsions composed by eudermic ingredients as glycerin, cocoa butter, almond oil and a variety of lipids, were enriched respectively with milk thistle dry extract (MT) or with a binary complex composed by MT and hydroxypropyl-β-cyclodextrin (HP) (1:4 w/w) correspondent to 1% (w/w) in sylimarine in order to obtain two different emulsions designed for the skin delivery and determine influence of hydroxypropyl-β-cyclodextrin on the extract delivery and permeation. Uv-vis spectrophotometric analyses demonstrated that phytocomplex formation influences the finding of MT after the complexation process and the in vitro antioxidant activity. Further in vitro and ex vivo experiments demonstrated that the penetration capability of MT from formulations is strictly influenced by the phytocomplex able to control MT permeation; moreover phytocomplex increases flavonoids stability during the in vitro tests. Additionally, in vivo studies showed that the penetration into the stratum corneum of the active ingredients is effectively achieved by the phytocomplex formation, in fact about 80% of MT is absorbed by the skin along 1h despite the 30% of MT not complexed absorbed during the same period.