Merck
  • Oridonin: targeting programmed cell death pathways as an anti-tumour agent.
Oridonin: targeting programmed cell death pathways as an anti-tumour agent.
Cell proliferation (2012-10-31)
Z Liu, L Ouyang, H Peng, W-Z Zhang
ABSTRACT
Oridonin, an active diterpenoid isolated from traditional Chinese herbal medicine, has drawn rising attention for its remarkable apoptosis- and autophagy-inducing activity and relevant molecular mechanisms in cancer therapy. Apoptosis is a well known type of cell death, whereas autophagy can play either pro-survival or pro-death roles in cancer cells. Accumulating evidence has recently revealed relationships between apoptosis and autophagy induced by oridonin; however, molecular mechanisms behind them remain to be discovered. In this review, we focus on highlighting updated research on oridonin-induced cell death signalling pathways implicated in apoptosis and autophagy, in many types of cancer. In addition, we further discuss cross-talk between apoptosis and autophagy induced by oridonin, in cancer. Taken together, these findings open new perspectives for further exploring oridonin as a potential anti-tumour agent targeting apoptosis and autophagy, in future anti-cancer therapeutics.
Sigma-Aldrich
Oridonin, ≥98% (HPLC), solid