Merck
  • IDH1 and IDH2 mutations in tumorigenesis: mechanistic insights and clinical perspectives.
IDH1 and IDH2 mutations in tumorigenesis: mechanistic insights and clinical perspectives.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-17)
Hui Yang, Dan Ye, Kun-Liang Guan, Yue Xiong
ABSTRACT
Genes encoding for isocitrate dehydrogenases 1 and 2, IDH1 and IDH2, are frequently mutated in multiple types of human cancer. Mutations targeting IDH1 and IDH2 result in simultaneous loss of their normal catalytic activity, the production of α-ketoglutarate (α-KG), and gain of a new function, the production of 2-hydroxyglutarate (2-HG). 2-HG is structurally similar to α-KG, and acts as an α-KG antagonist to competitively inhibit multiple α-KG-dependent dioxygenases, including both lysine histone demethylases and the ten-eleven translocation family of DNA hydroxylases. Abnormal histone and DNA methylation are emerging as a common feature of tumors with IDH1 and IDH2 mutations and may cause altered stem cell differentiation and eventual tumorigenesis. Therapeutically, unique features of IDH1 and IDH2 mutations make them good biomarkers and potential drug targets.
Sigma-Aldrich
IDH1 R132H (MRQ-67) Rabbit Monoclonal Antibody
Sigma-Aldrich
DL-α-Hydroxyglutaric acid disodium salt, ≥95.0% (GC)