Merck
  • Excitation functions for 7Be, 22,24Na production in Mg and Al by deuteron irradiations up to 50 MeV.
Excitation functions for 7Be, 22,24Na production in Mg and Al by deuteron irradiations up to 50 MeV.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2012-10-10)
A Hermanne, S Takács, F Tárkányi, R Adam-Rebeles, A Ignatyuk
ABSTRACT
New experimental data for production of (7)Be and (22,24)Na in deuteron irradiation of (nat)Mg and Al up to 50 MeV are presented. The induced activity, measured with HPGe spectroscopy, allows us to determine excitation functions of (nat)Mg(d,x) and (27)Al(d,x) reactions involved in the activation process with reference to (nat)Ti(d,x)(48)V monitor cross sections. A comparison with experimental literature values and results from updated theoretical codes is discussed. Thick target yields were derived from fits to our cross-sections and integrated personnel dose was calculated for different irradiation cycles and exposure scenarios around high power deuteron accelerator facilities.
Sigma-Aldrich
Beryllium, chunks, 99.9%