Merck
  • An efficient synthesis of 3-OBn-6β,14-epoxy-bridged opiates from naltrexone and identification of a related dual MOR inverse agonist/KOR agonist.
An efficient synthesis of 3-OBn-6β,14-epoxy-bridged opiates from naltrexone and identification of a related dual MOR inverse agonist/KOR agonist.
Bioorganic & medicinal chemistry letters (2012-07-10)
David J Martin, Paul E FitzMorris, Bo Li, Mario Ayestas, Ellicott J Sally, Christina M Dersch, Richard B Rothman, Amy M Deveau
ABSTRACT
In an effort to better understand the conformational preferences that inform the biological activity of naltrexone and related naltrexol derivatives, a new synthesis of the restricted analog 3-OBn-6β,14-epoxymorphinan 4 is described. 4 was synthesized starting from naltrexone in 50% overall yield, proceeding through the OBn-6α-triflate intermediate 8. Key steps to the synthesis include benzylation (96% yield), reduction (90% yield, α:β:3:2), followed by a one-pot triflation/displacement sequence (96% yield) to yield the desired bridged epoxy derivative 4. X-ray crystallographic analysis of intermediate 3-OBn-6α-naltrexol 7a supports population of the key boat conformation required for the epoxy ring closure. We also report that the 6β-mesylate 10-a high affinity opioid receptor ligand, the epimeric derivative of 11, and an analog of 12-functions as an inverse agonist at the mu opioid receptor using herkinorin pre-conditioned cells and an agonist at the kappa opioid receptor when evaluated in independent in vitro [(35)S]-GTP-γ-S assays.