Merck
  • [Effects of NaCl and Na2CO3 stresses on the growth and photosynthesis characteristics of Morus alba seedlings].
[Effects of NaCl and Na2CO3 stresses on the growth and photosynthesis characteristics of Morus alba seedlings].
Ying yong sheng tai xue bao = The journal of applied ecology (2012-06-23)
Hui-Hui Zhang, Xiu-Li Zhang, Xin Li, Jun-Nan Ding, Wen-Xu Zhu, Fei Qi, Ting Zhang, Ye Tian, Guang-Yu Sun
ABSTRACT
Taking 1-year old Morus alba variety 'Qinglong' seedlings as test materials, this paper studied their growth and photosynthetic characteristics under the stresses of different concentration neutral salt NaCl and alkali salt Na2CO3. Salt stresses decreased the plant height and the leaf number, biomass, and photosynthetic capacity of the seedlings markedly. With increasing concentration Na+, the leaf stomatal conductance, transpiration rate, net photosynthetic rate, actual photochemical efficiency, electron transport rate, and photochemical quenching (qP) decreased obviously, the energy dissipation rate increased, and the light use efficiency and photosynthetic capacity dropped down. At low concentrations Na+ (< 150 mmol x L(-1)), the seedlings growth and leaf photosynthetic capacity were slightly inhibited, and the adaptability of the seedlings to the salt stresses increased via the increase of root/shoot ratio. However, this protection mechanism was impaired by increasing salt concentration. Na2CO3 stress (Na+ concentration > 50 mmol x L(-)) had stronger inhibitory effects on the seedlings growth and leaf photosynthetic capacity, and the effect increased with increasing Na+ concentration. It was concluded that at Na+ concentration < 150 mmol x L(-1), the photosynthetic adaptability of M. alba to neutral salt stress was mainly dependent on the plant morphology and photosynthetic metabolism, but at Na+ concentration > 150 mmol x L(-1), the photosynthetic adaptability of M. alba to alkali salt stress was mainly dependent on the photosynthetic metabolism.
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7