Merck
Palladium-catalyzed N-arylation of 2-aminothiazoles.
Organic letters (2012-03-08)
Meredeth A McGowan, Jaclyn L Henderson, Stephen L Buchwald
ABSTRACT
A method for the Pd-catalyzed coupling of 2-aminothiazole derivatives with aryl bromides and triflates is described. Significantly, for this class of nucleophiles, the coupling exhibits a broad substrate scope and proceeds with a reasonable catalyst loading. Furthermore, an interesting effect of acetic acid as an additive is uncovered that facilitates catalyst activation.
Sigma-Aldrich
2-Aminothiazole, 97%