- Amphiphilic amylose-g-poly(meth)acrylate copolymers through "click" onto grafting method.
Amphiphilic amylose-g-poly(meth)acrylate copolymers through "click" onto grafting method.
Periodate oxidation and subsequent reductive amination with propargylamine was adopted for the controlled functionalization of amylose with alkyne groups, whereas ATRP polymerization was exploited to obtain end-(α)- or end-(ω)-azide functionalized poly(meth)acrylates to be used as "click" reagents in Cu(I) catalyzed azide-alkyne [3 + 2] dipolar cycloaddition. Amylose was effectively grafted with poly(n-butyl acrylate), poly(n-butyl methacrylate), poly(n-hexyl methacrylate), and poly(dimethylaminoethyl methacrylate) with this strategy. Their structure and composition were confirmed by FT-IR, NMR spectroscopies, and thermogravimetric analysis (TGA). Dynamic and static light scattering analyses, as well as TEM microscopy showed that the most amphiphilic among these hybrid graft copolymers self-assembled in water, yielding nanoparticles with ca. 30 nm diameter.