Merck
  • Mutagenic potentials of Amberlite XAD-2-resin extracts obtained from river and drinking waters in the Northwest district of Chiba, Japan.
Mutagenic potentials of Amberlite XAD-2-resin extracts obtained from river and drinking waters in the Northwest district of Chiba, Japan.
The Journal of toxicological sciences (2010-12-09)
Ayako Kanno, Iwaki Nishi, Tomohiro Kishi, Tsuyoshi Kawakami, Yasuo Takahashi, Sukeo Onodera
ABSTRACT
Amberlite XAD-2 resin extracts of river and drinking water sampled from the Northwest district of Chiba Prefecture in each month during the period from January to December 2008 were investigated to characterize and determine their mutagenic potentials and polycyclic aromatic hydrocarbon (PAH) levels. The extracts from the river water were shown to be mutagenic in Salmonella typhimurium TA98 (a flameshift mutagen) without S9 mix, with higher mutagenic responses in summer and early fall seasons. While the drinking water extracts exhibited weak mutagenicity in both the TA98 and TA100 strains (a base-pair substitution mutagen) without S9 mix, with high mutagenic responses in fall and early winter seasons. GC/MS determinations of the water concentrates showed some seasonal scatter in PAH levels in river water. In contrast, comparatively high concentrations of PAHs were observed for drinking water samples collected during warmer seasons. Statistical studies revealed that there is a lower correlation between the levels of flameshift mutagenicity and the concentrations of PAH in the river water concentrations, but a higher correlation between them in the drinking water samples.
Product Description
Supelco
Amberlite XAD®-2, pkg of 100 g
Supelco
Amberlite XAD®-2, pkg of 5 kg
Supelco
Amberlite XAD®-2, pkg of 25 kg
Supelco
Amberlite XAD®-2, pkg of 10 kg
Supelco
Amberlite XAD®-2, pkg of 500 g