Merck
  • Altered hapten recognition by two anti-digoxin hybridoma variants due to variable region point mutations.
Altered hapten recognition by two anti-digoxin hybridoma variants due to variable region point mutations.
The Journal of biological chemistry (1991-03-05)
J F Schildbach, D J Panka, D R Parks, G C Jager, J Novotny, L A Herzenberg, M Mudgett-Hunter, R E Bruccoleri, E Haber, M N Margolies
ABSTRACT
Two spontaneous variants of the murine anti-digoxin antibody-producing hybridoma cell line 26-10 were isolated by two-color fluorescence-activated cell sorting on the basis of altered hapten binding. The variable region sequences of the antibodies produced by the mutant lines revealed that each contains a single amino acid change in the heavy chain second complementarity determining region. A Tyr to His change at position 50 leads to a 40-fold reduction in affinity for digoxin. A Ser to Phe mutation at position 52 results in a 300-fold reduction in affinity for digoxin. A competition assay involving 33 digoxin analogues was used to examine the specificity of hapten binding of 26-10 and the two mutant antibodies. The position 50 mutant has a distinct specificity change; it exhibits a preference for digoxin congeners containing a hydroxyl group at the steroid 12 position, whereas the 26-10 parent does not. The affinities of all three antibodies for hapten are progressively lowered by substitutions of increasing size at the digoxin steroid D ring 16 position. Although 26-10 binds digoxin and its genin form equally, 12 and 16 steroid position substitutions which lower affinity also confer a preference for a sugar at the steroid 3 position. These results suggest that position 50 contributes to specificity of the antibody and that alterations of the hapten can lead to differences in recognition, possibly through a shift in hapten orientation within the binding site.
Sigma-Aldrich
Gitoxin, ≥99% (HPLC)