Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Genetic interactions at the FLA10 locus: suppressors and synthetic phenotypes that affect the cell cycle and flagellar function in Chlamydomonas reinhardtii.
Genetic interactions at the FLA10 locus: suppressors and synthetic phenotypes that affect the cell cycle and flagellar function in Chlamydomonas reinhardtii.
Genetics (1991-07-01)
F G Lux, S K Dutcher
ABSTRACT
Through the isolation of suppressors of temperature-sensitive flagellar assembly mutations at the FLA10 locus of Chlamydomonas reinhardtii, we have identified six other genes involved in flagellar assembly. Mutations at these suppressor loci, termed SUF1-SUF6, display allele specificity with respect to which fla10- mutant alleles they suppress. An additional mutation, apm1-122, which confers resistance to the plant herbicides amiprophos-methyl and oryzalin, was also found to interact with mutations at the FLA10 locus. The apm1-122 mutation in combination with three fla10- mutant alleles results in synthetic cold-sensitive cell division defects, and in combination with an additional pseudo-wild-type fla10- allele yields a synthetic temperature-sensitive flagellar motility phenotype. Based upon the genetic interactions of these loci, we propose that the FLA10 gene product interacts with multiple components of the flagellar apparatus and plays a role both in flagellar assembly and in the cell cycle.
Sigma-Aldrich
Amiprofos methyl, PESTANAL®, analytical standard