Merck
  • Probing lipid vesicles by bimolecular association and dissociation trajectories of single molecules.
Probing lipid vesicles by bimolecular association and dissociation trajectories of single molecules.
Journal of the American Chemical Society (2006-04-06)
Feng Gao, Erwen Mei, Manho Lim, Robin M Hochstrasser
ABSTRACT
Vesicles prepared by DMPC (1,2-dimyristoyl-sn-glycero-3-phosphocholine) and SOPC (1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine) lipid molecules having sizes smaller than the diffraction-limited focused laser beam have been used to confine single molecules in the laser focus. The confinement of single molecules in a volume smaller than the focused laser beam leads to a Gaussian distribution of single molecule fluorescence intensity. The interactions of single Nile Red molecules with DMPC and SOPC lipid bilayers were studied by single molecule fluorescence confocal microscopy. Nile Red molecules were observed to associate with and dissociate from individual DMPC and SOPC vesicles adsorbed on a glass surface, generating on-and-off fluctuations in a fluorescence signal representing a very low noise two-state trajectory. Off-time statistics were used to investigate the mean radius of the vesicles and the size distribution functions. The means of the on-time distributions of Nile Red in DMPC and SOPC vesicles were significantly different. The association and dissociation reactions of single Nile Red molecules with a vesicle have been studied. Features of the bimolecular interaction between the probe Nile Red and the vesicle were evaluated from the uncorrelated mean on-time and vesicle radius distributions, and the linear Nile Red concentration dependence of the mean off-time. Nile Red is shown to be a useful probe of the structural fluctuations and heterogeneity of these membrane structures, and it is a useful model with which to directly study a diffusion-influenced reversible bimolecular reaction.