- Successive optical resolution of two compounds by one enantiopure compound.
Successive optical resolution of two compounds by one enantiopure compound.
Chemical communications (Cambridge, England) (2006-03-04)
Yoshitane Imai, Masatoshi Takeshita, Tomohiro Sato, Reiko Kuroda
PMID16514442
ABSTRACT
By using (1R,2R)-1,2-diphenylethylenediamine as a single enantiopure compound, we achieved a novel successive optical resolution of more than one kind of racemic compound through supramolecular crystallization.
MATERIALS