Merck
  • Practical enantioselective synthesis of beta-substituted-beta-amino esters.
Practical enantioselective synthesis of beta-substituted-beta-amino esters.
The Journal of organic chemistry (2005-07-02)
Alok K Awasthi, Mark L Boys, Kimberly J Cain-Janicki, Pierre-Jean Colson, Wendel W Doubleday, Joseph E Duran, Payman N Farid
ABSTRACT
[reaction: see text] A practical, large-scale synthesis of a beta-amino ester 1 was developed. A chiral imine derived from (S)-phenylglycinol and 3-trimethylsilylpropanal was coupled with the Reformatsky reagent 3 with high diastereoselectivity (de > 98%) to give (SS)-4a as the major isomer. The amino alcohol residue of the coupling product 4 was oxidatively cleaved with sodium periodate in the presence of methylamine. An unusual selective oxidative cleavage of the (SS)-isomer was observed and the imine 6 was obtained with ee > 99% while the (RS)-4b isomer was not cleaved. Reaction with p-toluenesulfonic acid monohydrate allowed for the hydrolysis of the imine and the isolation of the amine as its salt. The title compound 1 was then obtained by transesterification, desilylation, and hydrochloride salt formation in a one-pot process. The method was successfully applied toward the synthesis of a wide variety of beta-amino esters.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%