Merck
  • Mechanisms involved in the contraction of intrahepatic portal vein branches by clomipramine and oxethazaine in isolated perfused rat livers.
Mechanisms involved in the contraction of intrahepatic portal vein branches by clomipramine and oxethazaine in isolated perfused rat livers.
Journal of pharmacological sciences (2005-06-09)
Yasusuke Masuda, Tomoko Edo
ABSTRACT
Clomipramine (CLM) and oxethazaine (OXZ) were previously reported to increase portal pressure by contracting portal vein branches (PVBs) in isolated perfused rat liver. In the present study, to characterize the contractile mechanisms, the effects of Y27632, HA1077, staurosporine, papaverine, SKF96365, and sulindac sulfide on the portal pressure increase induced by CLM and OXZ were examined comparatively with those induced by endothelin-1. The results suggest that 1) intrahepatic PVBs employ a Rho-kinase-dependent pathway for sustained contraction, 2) CLM contracts PVBs by activating a Rho-kinase pathway and Ca(2+)-channels, and 3) OXZ acts primarily by promoting Ca(2+) entry through its ionophore-like action.
Product Description
Supelco
Oxethazaine, analytical standard