Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Hyaluronan conformations on surfaces: effect of surface charge and hydrophobicity.
Hyaluronan conformations on surfaces: effect of surface charge and hydrophobicity.
Carbohydrate research (2005-03-23)
Chiara Spagnoli, Alexander Korniakov, Abraham Ulman, Endre A Balazs, Yuri L Lyubchenko, Mary K Cowman
ABSTRACT
Extended, relaxed, condensed, and interacting forms of the polysaccharide hyaluronan have been observed by atomic force microscopy (AFM). The types of images obtained depend on the properties of the surfaces used. We have investigated several different surface conditions for HA imaging, including unmodified mica, mica chemically modified with two different kinds of amino-terminated silanes (3-aminopropyltriethoxysilane and N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride), and highly oriented pyrolytic graphite. We found the degree of HA molecular extension or condensation to be variable, and the number of bound chains per unit area was low, for all of the mica-based surfaces. HA was more easily imaged on graphite, a hydrophobic surface. Chains were frequently observed in high degrees of extension, maintained by favorable interaction with the surface after molecular combing. This observation suggests that the HA macromolecule interacts with graphite through hydrophobic patches along its surface. AFM studies of HA behavior on differing surfaces under well-controlled environmental conditions provides useful insight into the variety of conformations and interactions likely to be found under differing physiological conditions.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl-N,N,N-trimethylammonium-15N chloride, 99 atom % 15N, 97% (CP)