Merck
  • Antidiabetic effect of a nitrosamine-free dephostatin analogue, methoxime-3,4-dephostatin, in db/db mice.
Antidiabetic effect of a nitrosamine-free dephostatin analogue, methoxime-3,4-dephostatin, in db/db mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2002-07-12)
A Hiroki, H Hatakeyama, M Kawakami, T Watanabe, I Takei, K Umezawa
ABSTRACT
Et-3,4-dephostatin, a protein-tyrosine phosphatase (PTPase) inhibitor, potentiates insulin-dependent signal transduction and shows an antidiabetic effect in mice. However, it contains a nitrosamine moiety that is often mutagenic and carcinogenic. Therefore, we previously designed and synthesized methoxime-3,4-dephostatin as a nitrosamine-free analogue of dephostatin. In the present paper, we studied in situ and in vivo antidiabetic effects of this PTPase inhibitor. Methoxime-3,4-dephostatin induced 2-deoxyglucose transport by mouse 3T3-L1 adipocytes and rat L6 myocytes without insulin. It also inhibited glucagon-induced glucose release from primary culture rat hepatocytes. When hepatocytes were prepared from starved rats, methoxime-3,4-dephostatin did not inhibit the release of glucose, indicating that the chemical may act on glycogenolysis. Oral administration of methoxime-3,4-dephostatin for 3-7 days inhibited the increase in the blood glucose level in type-2 diabetes model db/db mice. It also decreased food and water intakes of mice, but showed no liver or blood toxicity.