Merck
  • The antioxidant activity of phloretin: the disclosure of a new antioxidant pharmacophore in flavonoids.
The antioxidant activity of phloretin: the disclosure of a new antioxidant pharmacophore in flavonoids.
Biochemical and biophysical research communications (2002-06-27)
Bashir M Rezk, Guido R M M Haenen, Wim J F van der Vijgh, Aalt Bast
ABSTRACT
Phloretin is a dihydrochalcone flavonoid that displays a potent antioxidant activity in peroxynitrite scavenging and the inhibition of lipid peroxidation. Comparison with structurally related compounds revealed that the antioxidant pharmacophore of phloretin is 2,6-dihydroxyacetophenone. The potent activity of 2,6-dihydroxyacetophenone is due to stabilisation of its radical via tautomerisation. The antioxidant pharmacophore in the dihydrochalcone phloretin, i.e., the 2,6-dihydroxyacetophenone group, is different from the antioxidant pharmacophores previously reported in flavonoids.
Product Description
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
Phloretin, ≥99%