Merck
[A clinicopathological study of pancreatoblastoma].
Zhonghua bing li xue za zhi = Chinese journal of pathology (2002-03-01)
L He, P Li, S Liu, L Wang, N Liu, X Du
ABSTRACT
To investigate the clinicopathological, immunohistochemical features and the histogenesis of pancreatoblastoma (PB) in 14 pediatric cases, including 9 male and 5 female patients, their age ranged from 1.5 to 8 years with a mean of 4.4 years. Routine pathological, histochemical, and immunohistochemical methods were utilized to analyse the PB specimens. Tumors of 6 patients had metastasized to the liver, spleen and lymph nodes, 8 patients were alive 10 months to 105 months after diagnosis. Histologically, the tumors were composed of dense epithelial elements separated by fibro-stromal tissue, resulting in a nesting or organoid pattern. Immunohistochemically and histochemically, the tumors exhibited acinar, endocrine and ductal differentiation. The positive rates for were: CK 100%, EMA 86%, NSE 79%, CEA 71%, SYN 64%, CgA 43%, alpha1-AT 57%, PAS 100% and mucicarmine 57%. PB is the most common malignant pancreatic neoplasm of childhood. PB arises from primitive pluripotential cells capable of differentiating into 3 pancreatic cell types: acinar, endocrine and ductal. The prognosis is better than pancreatic carcinoma in adults.
Sigma-Aldrich
Mayer′s Mucicarmine Stain Solution, for microscopy