Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Sulphonation of 2,6-dimethoxynaphthalene. Reaction mechanisms and products.
Sulphonation of 2,6-dimethoxynaphthalene. Reaction mechanisms and products.
Acta poloniae pharmaceutica (2000-12-29)
A Cisak, D Kusztal, E Brzezińska
ABSTRACT
Protonation and sulphonation mechanism of 2,6-DMON is described. The products of their sulphonation, the sodium and barium salts of 2,6-DMON-4-monosulphonic acid and of 2,6-DMON-4,7-disulphonic acid have been prepared and characterized.