Merck
  • Allylmalonamide as a bivalent linker: synthesis of biantennary GM3-saccharide--keyhole limpet hemocyanin glycoconjugate and the immune response in mice.
Allylmalonamide as a bivalent linker: synthesis of biantennary GM3-saccharide--keyhole limpet hemocyanin glycoconjugate and the immune response in mice.
Glycoconjugate journal (2000-05-18)
W Zou, M Abraham, M Gilbert, W W Wakarchuk, H J Jennings
ABSTRACT
A biantennary GM3-saccharide (sialyllactoside) derivative (4) was constructed using allylmalonic acid as a bivalent linker, both carboxylic acids of which were condensed with 3-aminopropyl lactoside (2) prior to enzymatic sialylation with a fusion enzyme. While ozonolysis of its allyl group generated a saccharide having a terminal aldehyde (6), we were unable to couple 6 directly to protein by reductive amination. However, extension of the spacer by means of introducing a maleimide group to 6 through its aldehyde group to give 7 enabled the latter to be successfully coupled to thiolated proteins. The average ratios of saccharide to protein were observed to be 35 in KLH conjugate (13) and 9-12 in HSA conjugates (14 and 15). The antisera obtained by immunizing mice with the biantennary sialyllactoside-KLH conjugate (13) together with MPL adjuvant were analyzed by ELISA. Using several structurally related saccharide-HSA conjugates as screening antigens, it was concluded that anti-sialyllactoside antibodies, both IgG and IgM, were effectively raised. This was further supported by competitive inhibition experiments using lactoside (1), sialyllactoside (8) and biantennary sialyllactoside (4) as inhibitors.
Sigma-Aldrich
Allylmalonic acid, ≥98.0% (T)