Merck
  • Highly conductive and stretchable polymer composites based on graphene/MWCNT network.
Highly conductive and stretchable polymer composites based on graphene/MWCNT network.
Chemical communications (Cambridge, England) (2013-01-22)
Mengting Chen, Tao Tao, Ling Zhang, Wei Gao, Chunzhong Li
ABSTRACT
We have manufactured a highly conductive and stretchable composite by backfilling a preformed graphene/MWCNT aerogel with poly(dimethylsiloxane) (PDMS). The electrical conductivity of our product can reach 2.8 S cm(-1) with only 1.3 wt% graphene/MWCNT loading, and remains constant after 100 times repeated stretching by 20% and 5000 times bending.
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)