Merck
  • On-step synthesis of a bifunctional BINOL ligand for the highly enantioselective cyanation of aliphatic aldehydes.
On-step synthesis of a bifunctional BINOL ligand for the highly enantioselective cyanation of aliphatic aldehydes.
Organic letters (2005-06-04)
Ying-Chuan Qin, Lan Liu, Lin Pu
ABSTRACT
[reaction: see text] An efficient one-step synthesis of the optically active bifunctional BINOL ligand (S)-3 has been developed. It was found that (S)-3 in combination with Me2AlCl is a highly enantioselective catalyst for the addition of TMSCN to aliphatic aldehydes of diverse structures and is also among the most practical ones. A remarkable positive nonlinear effect was found for this chiral ligand.
Sigma-Aldrich
Trimethylsilyl cyanide, technical, ≥95% (GC)
Sigma-Aldrich
Trimethylsilyl cyanide, 98%