Merck
Biodegradation of dimethylterephthalate by Comamonas acidovorans D-4.
Indian journal of experimental biology (1998-10-01)
D S Patel, A J Desai, J D Desai
ABSTRACT
Comamonas acidovorans D-4, capable of utilizing dimethylterephthalate (DMT) as the sole carbon source, was isolated from the activated sludge of petrochemical wastewater treatment plant. Almost complete utilization of as high as 0.5% (w/v) DMT was observed in 72 hr. Growth kinetics followed a parallel relation between the growth, DMT utilization and cell associated esterase activity. A cell free broth of DMT grown cells showed an extracellular esterase activity. During the DMT degradation an extracellular accumulation of terephthalic acid was found. Although, C. acidovorans grew on a number of phthalate esters and phthalic acids as the sole carbon source, growth was significantly high on phthalic acids. The potential of this organism in petrochemical pollution abatement is discussed.
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)