Merck
  • Selective Clara cell injury in mouse lung following acute administration of coumarin.
Selective Clara cell injury in mouse lung following acute administration of coumarin.
Toxicology and applied pharmacology (1998-08-26)
S L Born, A S Fix, D Caudill, L D Lehman-McKeeman
ABSTRACT
Coumarin is a known hepatotoxicant in laboratory animals, particularly rats. However, the mouse lung was identified as a major target organ in a chronic bioassay, with an oral gavage dosage of 200 mg/kg coumarin increasing the incidence of alveolar/bronchiolar adenomas and carcinomas. The purpose of the present work was to determine whether coumarin was acutely toxic in the mouse and rat lung. Male and female B6C3F1 mice were dosed orally by gavage with coumarin at 0, 10, 20, 50, 100, 150, and 200 mg/kg and lung toxicity was determined 24 h later by histological evaluation. The results indicated that coumarin dosages >/= 150 mg/kg caused selective injury to Clara cells in the distal bronchiolar epithelium. The time course of this injury was studied from 6 h to 7 days after a single dosage of coumarin (200 mg/kg). At 12 h after dosing, Clara cell swelling was apparent along with the onset of necrosis and bronchiolar epithelial disorganization. At 24-48 h, necrotic Clara cells were observed sloughed into the lumens of the terminal bronchioles, with concomitant thinning of the epithelium and flattening of the remaining ciliated cells. By 72-96 h, there was epithelial hypertrophy and hyperplasia, and by 7 days after dosing, the Clara cells had regenerated and the bronchiolar epithelial architecture appeared nearly normal. Unlike the mouse, oral administration of coumarin (200 mg/kg) caused severe hepatotoxicity in male F344 rats, seen histologically as centrilobular necrosis and associated with increases, up to 140-fold, in serum ALT, AST, and SDH levels. Clara cell toxicity was not observed in the distal bronchioles of treated rats. However, in the upper airways, coumarin treatment produced generalized epithelial necrosis involving both ciliated and nonciliated cells. 3,4-Dihydrocoumarin (DHC), which is not a mouse lung carcinogen, did not cause Clara cell injury when dosed to mice at 800 mg/kg. This finding suggests, because DHC lacks a 3,4-double bond, that bioactivation of coumarin to a 3,4-epoxide intermediate may contribute to mouse lung Clara cell toxicity. Collectively, the results indicate that coumarin is a Clara cell toxicant and establish the mouse lung as a target organ for coumarin toxicity. These new findings lay the foundation for studies to determine the mechanisms of coumarin-induced toxicity and carcincogenicity and to define the relevance of these effects to humans.
Sigma-Aldrich
Dihydrocoumarin, ≥99%, FCC, FG
Sigma-Aldrich
Dihydrocoumarin, 99%